Assay Details
| Gene Symbol: TAFA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077000.2 | NP_001070468.1 | 3 - 4 | 453 | 98 | |
| GenBank mRNA | BC103140.1 | - | 3 - 4 | 454 | 98 | |
More Information
Set Membership: |
|

TAFA5
Genomic Map