Assay Details
| Gene Symbol: MPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077042.2 | NP_001070510.1 | 4 - 5 | 438 | 85 | |
| GenBank mRNA | BC102377.1 | - | 4 - 5 | 439 | 85 | |
More Information
Set Membership: |
|

MPC1
Genomic Map