Assay Details
| Gene Symbol: SERPINA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173882.2 | NP_776307.1 | 1 - 2 | 24 | 53 | |
| GenBank mRNA | BC102730.1 | - | 1 - 2 | 42 | 53 | |
| X63129.1 | - | 25 | 53 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102730.1, NM_173882.2, X63129.1 transcripts.
Set Membership: |
|

SERPINA1
Genomic Map