Assay Details
| Gene Symbol: ADA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173887.2 | NP_776312.1 | 9 - 10 | 843 | 57 | |
| GenBank mRNA | AF280603.1 | - | 9 - 10 | 843 | 57 | |
| BC146169.1 | - | 9 - 10 | 929 | 57 | ||
More Information
Set Membership: |
|

ADA
Genomic Map