Assay Details
| Gene Symbol: FGG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173911.2 | NP_776336.1 | 6 - 7 | 676 | 106 | |
| GenBank mRNA | BC102629.1 | - | 6 - 7 | 702 | 106 | |
| X15556.1 | - | 676 | 106 | |||
More Information
Set Membership: |
|

FGG
Genomic Map