Assay Details
| Gene Symbol: MIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173937.2 | NP_776362.1 | 1 - 2 | 412 | 57 | |
| GenBank mRNA | K02818.1 | - | 1 - 2 | 412 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173937.2 | NP_776362.1 | 1 - 2 | 412 | 57 | |
| GenBank mRNA | K02818.1 | - | 1 - 2 | 412 | 57 | |
Set Membership: |
|