Assay Details
Gene Symbol: PRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173953.2 | NP_776378.2 | 4 - 5 | 569 | 67 | |
GenBank mRNA | AH001155.2 | - | 4 - 5 | 574 | 67 | |
BC148124.1 | - | 4 - 5 | 553 | 67 | ||
M25007.1 | - | 1 - 2 | 119 | 67 | ||
MH477634.1 | - | 2 - 3 | 67 | |||
V00112.1 | - | 569 | 67 | |||
X14321.1 | - | 217 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
