Assay Details
Gene Symbol: AMBN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173988.2 | NP_776413.1 | 3 - 4 | 233 | 69 | |
GenBank mRNA | AF157019.1 | - | 3 - 4 | 233 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
