Assay Details
Gene Symbol: CST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174029.1 | NP_776454.1 | 2 - 3 | 418 | 70 | |
GenBank mRNA | AB098964.1 | - | 2 - 3 | 399 | 70 | |
AB099041.1 | - | 2 - 3 | 399 | 70 | ||
BC109629.1 | - | 2 - 3 | 419 | 70 | ||
Y10811.1 | - | 418 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
