Assay Details
| Gene Symbol: MOCOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174081.2 | NP_776506.1 | 6 - 7 | 1507 | 59 | |
| GenBank mRNA | AB036422.1 | - | 6 - 7 | 1507 | 59 | |
More Information
Set Membership: |
|

MOCOS
Genomic Map