Assay Details
| Gene Symbol: HPSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174082.2 | NP_776507.1 | 8 - 9 | 1122 | 59 | |
| GenBank mRNA | AF281160.2 | - | 8 - 9 | 1122 | 59 | |
More Information
Set Membership: |
|

HPSE
Genomic Map