Assay Details
| Gene Symbol: IL1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174093.1 | NP_776518.1 | 2 - 3 | 119 | 71 | |
| GenBank mRNA | EU276067.1 | - | 1 - 2 | 74 | 71 | |
| M35589.1 | - | 2 - 3 | 90 | 71 | ||
| M37211.1 | - | 2 - 3 | 119 | 71 | ||
| X12498.1 | - | 1 | 90 | 71 | ||
More Information
Set Membership: |
|

IL1B
Genomic Map