Assay Details
Gene Symbol: LIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174102.2 | NP_776527.1 | 1 - 2 | 54 | 56 | |
GenBank mRNA | L04188.1 | - | 1 - 2 | 54 | 56 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174102.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
