Assay Details
Gene Symbol: MC4R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174110.1 | NP_776535.1 | 1 | 1452 | 149 | |
GenBank mRNA | AF265221.1 | - | 1 | 1452 | 149 | |
BC148892.1 | - | 1 | 1399 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
