Assay Details
| Gene Symbol: NPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174127.2 | NP_776552.1 | 3 - 4 | 1513 | 63 | |
| GenBank mRNA | BC151352.1 | - | 3 - 4 | 1069 | 63 | |
| D13508.1 | - | 3 - 4 | 1048 | 63 | ||
| J03876.1 | - | 3 - 4 | 1513 | 63 | ||
More Information
Set Membership: |
|

NPR3
Genomic Map