Assay Details
| Gene Symbol: POMC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174151.1 | NP_776576.1 | 2 - 3 | 244 | 62 | |
| GenBank mRNA | BC122728.1 | - | 1 - 2 | 141 | 62 | |
| M25587.1 | - | 2 - 3 | 244 | 62 | ||
| MK309341.1 | - | 2 - 3 | 167 | 62 | ||
| V00107.1 | - | 244 | 62 | |||
More Information
Set Membership: |
|

POMC
Genomic Map