Assay Details
Gene Symbol: PRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174160.2 | NP_776585.1 | 1 - 2 | 238 | 96 | |
GenBank mRNA | M27240.1 | - | 1 - 2 | 238 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
