Assay Details
| Gene Symbol: GRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174173.2 | NP_776598.1 | 5 - 6 | 1389 | 66 | |
| GenBank mRNA | M73836.1 | - | 5 - 6 | 1389 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRK1| Gene Symbol: GRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174173.2 | NP_776598.1 | 5 - 6 | 1389 | 66 | |
| GenBank mRNA | M73836.1 | - | 5 - 6 | 1389 | 66 | |
Set Membership: |
|