Assay Details
| Gene Symbol: TSFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174204.3 | NP_776629.1 | 1 - 2 | 119 | 89 | |
| GenBank mRNA | BC110136.1 | - | 1 - 2 | 125 | 89 | |
| BC149988.1 | - | 1 - 2 | 144 | 89 | ||
| L37935.1 | - | 1 - 2 | 119 | 89 | ||
More Information
Set Membership: |
|

TSFM
Genomic Map