Assay Details
Gene Symbol: TXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174208.2 | NP_776633.1 | 2 - 3 | 330 | 62 | |
GenBank mRNA | BC112876.1 | - | 2 - 3 | 360 | 62 | |
D87741.1 | - | 2 - 3 | 330 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
