Assay Details
| Gene Symbol: UQCR11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174215.2 | NP_776640.1 | 2 - 3 | 210 | 81 | |
| GenBank mRNA | BC102080.1 | - | 2 - 3 | 236 | 81 | |
| BC126774.1 | - | 2 - 3 | 214 | 81 | ||
| D55637.1 | - | 2 - 3 | 210 | 81 | ||
More Information
Set Membership: |
|

UQCR11
Genomic Map