Assay Details
| Gene Symbol: ATP6V1G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174245.2 | NP_776670.1 | 2 - 3 | 331 | 74 | |
| GenBank mRNA | BC110010.1 | - | 2 - 3 | 287 | 74 | |
| S82464.1 | - | 2 - 3 | 331 | 74 | ||
More Information
Set Membership: |
|

ATP6V1G1
Genomic Map