Assay Details
Gene Symbol: CLIC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174276.3 | NP_776701.1 | 3 - 4 | 1005 | 72 | |
GenBank mRNA | AF109199.1 | - | 3 - 4 | 899 | 72 | |
L16547.1 | - | 3 - 4 | 1006 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
