Assay Details
| Gene Symbol: GNAI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174324.2 | NP_776749.1 | 7 - 8 | 992 | 67 | |
| GenBank mRNA | BC105419.1 | - | 8 - 9 | 1034 | 67 | |
| X03642.1 | - | 992 | 67 | |||
More Information
Set Membership: |
|

GNAI1
Genomic Map