Assay Details
Gene Symbol: ICAM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174349.1 | NP_776774.1 | 6 - 7 | 1541 | 92 | |
GenBank mRNA | L41844.1 | - | 6 - 7 | 1541 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
