Assay Details
| Gene Symbol: IL2RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174358.2 | NP_776783.1 | 4 - 5 | 777 | 111 | |
| GenBank mRNA | BC133546.1 | - | 4 - 5 | 745 | 111 | |
| M20818.1 | - | 4 - 5 | 777 | 111 | ||
More Information
Set Membership: |
|

IL2RA
Genomic Map