Assay Details
| Gene Symbol: NPY2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174401.2 | NP_776826.1 | 1 - 2 | 124 | 60 | |
| GenBank mRNA | U50144.1 | - | 1 - 2 | 124 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_174401.2 transcripts.
Set Membership: |
|

NPY2R
Genomic Map