Assay Details
Gene Symbol: P2RY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174410.3 | NP_776835.1 | 1 | 1540 | 112 | |
GenBank mRNA | U34041.1 | - | 1 | 1193 | 112 | |
X87628.1 | - | 14 | 1548 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
