Assay Details
| Gene Symbol: PDE6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174420.2 | NP_776845.1 | 1 - 2 | 213 | 63 | |
| GenBank mRNA | U65073.1 | - | 1 - 2 | 213 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDE6D| Gene Symbol: PDE6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174420.2 | NP_776845.1 | 1 - 2 | 213 | 63 | |
| GenBank mRNA | U65073.1 | - | 1 - 2 | 213 | 63 | |
Set Membership: |
|