Assay Details
| Gene Symbol: PDE6G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174421.2 | NP_776846.1 | 1 - 2 | 194 | 72 | |
| GenBank mRNA | X04270.1 | - | 194 | 72 | ||
| X04823.1 | - | 194 | 72 | |||
More Information
Set Membership: |
|

PDE6G
Genomic Map