Assay Details
Gene Symbol: PDE6G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174421.2 | NP_776846.1 | 2 - 3 | 236 | 62 | |
GenBank mRNA | X04270.1 | - | 236 | 62 | ||
X04823.1 | - | 236 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
