Assay Details
| Gene Symbol: PPM1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302772.1 | NP_001289701.1 | 2 - 3 | 1070 | 81 | |
| NM_174429.3 | NP_776854.1 | 1 - 2 | 860 | 81 | ||
| GenBank mRNA | AJ005457.1 | - | 2 - 3 | 894 | 81 | |
| BC112558.1 | - | 2 - 3 | 1069 | 81 | ||
More Information
Set Membership: |
|

PPM1A
Genomic Map