Assay Details
| Gene Symbol: RPL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174455.3 | NP_776880.1 | 4 - 5 | 367 | 100 | |
| GenBank mRNA | BC102108.1 | - | 4 - 5 | 366 | 100 | |
More Information
Set Membership: |
|

RPL24
Genomic Map