Assay Details
Gene Symbol: SSTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174467.1 | NP_776892.1 | 1 | 1049 | 80 | |
GenBank mRNA | L06613.1 | - | 1 | 1049 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
