Assay Details
| Gene Symbol: VAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174483.2 | NP_776908.1 | 4 - 5 | 483 | 65 | |
| GenBank mRNA | X76199.1 | - | 483 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VAMP2| Gene Symbol: VAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174483.2 | NP_776908.1 | 4 - 5 | 483 | 65 | |
| GenBank mRNA | X76199.1 | - | 483 | 65 | ||
Set Membership: |
|