Assay Details
| Gene Symbol: ADAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174513.2 | NP_776938.1 | 6 - 7 | 692 | 63 | |
| GenBank mRNA | D89940.1 | - | 6 - 7 | 692 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADAP1| Gene Symbol: ADAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174513.2 | NP_776938.1 | 6 - 7 | 692 | 63 | |
| GenBank mRNA | D89940.1 | - | 6 - 7 | 692 | 63 | |
Set Membership: |
|