Assay Details
| Gene Symbol: GABRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174542.2 | NP_776967.1 | 5 - 6 | 612 | 68 | |
| GenBank mRNA | BC122761.1 | - | 5 - 6 | 741 | 68 | |
| X12362.1 | - | 612 | 68 | |||
More Information
Set Membership: |
|

GABRA3
Genomic Map