Assay Details
Gene Symbol: OPN1LW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174566.1 | NP_776991.1 | 1 - 2 | 110 | 70 | |
GenBank mRNA | AF280398.1 | - | 1 - 2 | 110 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
