Assay Details
| Gene Symbol: OPN1LW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174566.1 | NP_776991.1 | 5 - 6 | 982 | 92 | |
| GenBank mRNA | AF280398.1 | - | 5 - 6 | 982 | 92 | |
More Information
Set Membership: |
|

OPN1LW
Genomic Map