Assay Details
Gene Symbol: PABPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174569.2 | NP_776994.1 | 3 - 4 | 2631 | 67 | |
GenBank mRNA | BC133468.1 | - | 3 - 4 | 550 | 67 | |
BC133551.1 | - | 4 - 5 | 683 | 67 | ||
BC133558.1 | - | 3 - 4 | 541 | 67 | ||
X89969.1 | - | 2631 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
