Assay Details
| Gene Symbol: PRKACA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174584.2 | NP_777009.1 | 3 - 4 | 239 | 63 | |
| GenBank mRNA | X67154.1 | - | 239 | 63 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRKACA| Gene Symbol: PRKACA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174584.2 | NP_777009.1 | 3 - 4 | 239 | 63 | |
| GenBank mRNA | X67154.1 | - | 239 | 63 | ||
Set Membership: |
|