Assay Details
| Gene Symbol: SLC2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174602.2 | NP_777027.1 | 7 - 8 | 1144 | 55 | |
| GenBank mRNA | BC119940.1 | - | 7 - 8 | 1133 | 55 | |
| BC151708.1 | - | 6 - 7 | 985 | 55 | ||
| BT029806.1 | - | 7 - 8 | 1132 | 55 | ||
| M60448.1 | - | 7 - 8 | 1144 | 55 | ||
More Information
Set Membership: |
|

SLC2A1
Genomic Map