Assay Details
| Gene Symbol: PRDX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174643.1 | NP_777068.1 | 2 - 3 | 291 | 111 | |
| GenBank mRNA | AF080228.1 | - | 2 - 3 | 315 | 111 | |
| AF090194.1 | - | 2 - 3 | 291 | 111 | ||
| AJ243848.1 | - | 2 - 3 | 301 | 111 | ||
| BC102172.1 | - | 2 - 3 | 316 | 111 | ||
| BT020967.1 | - | 2 - 3 | 305 | 111 | ||
More Information
Set Membership: |
|

PRDX6
Genomic Map