Assay Details
Gene Symbol: IDH3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174644.2 | NP_777069.1 | 5 - 6 | 517 | 65 | |
GenBank mRNA | BC118259.1 | - | 5 - 6 | 481 | 65 | |
U07980.1 | - | 5 - 6 | 517 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
