Assay Details
| Gene Symbol: PLA2G1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174646.3 | NP_777071.2 | 3 - 4 | 421 | 88 | |
| GenBank mRNA | DN543510.1 | - | 3 - 4 | 421 | 88 | |
| Y00120.1 | - | 346 | 88 | |||
More Information
Set Membership: |
|

PLA2G1B
Genomic Map