Assay Details
| Gene Symbol: S100A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174650.1 | NP_777075.1 | 1 - 2 | 45 | 67 | |
| GenBank mRNA | AY911333.1 | - | 1 - 2 | 56 | 67 | |
| BC102207.1 | - | 1 - 2 | 62 | 67 | ||
| M16464.1 | - | 1 - 2 | 45 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102207.1, NM_174650.1 transcripts.
Set Membership: |
|

S100A10
Genomic Map