Assay Details
| Gene Symbol: S100A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174650.1 | NP_777075.1 | 2 - 3 | 195 | 98 | |
| GenBank mRNA | AB098817.1 | - | 1 - 2 | 405 | 98 | |
| AY911333.1 | - | 2 - 3 | 206 | 98 | ||
| BC102207.1 | - | 2 - 3 | 212 | 98 | ||
| M16464.1 | - | 2 - 3 | 195 | 98 | ||
More Information
Set Membership: |
|

S100A10
Genomic Map