Assay Details
Gene Symbol: SLC25A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174658.2 | NP_777083.1 | 2 - 3 | 614 | 83 | |
GenBank mRNA | BC102994.1 | - | 2 - 3 | 711 | 83 | |
M24102.1 | - | 2 - 3 | 616 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
