Assay Details
| Gene Symbol: NDUFB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174667.3 | NP_777092.1 | 3 - 4 | 345 | 70 | |
| GenBank mRNA | AY911318.1 | - | 3 - 4 | 325 | 70 | |
| BC103425.1 | - | 1 | 325 | 70 | ||
| BC151404.1 | - | 1 | 326 | 70 | ||
| X63209.1 | - | 346 | 70 | |||
More Information
Set Membership: |
|

NDUFB8
Genomic Map