Assay Details
Gene Symbol: CELA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174714.2 | NP_777139.1 | 1 - 2 | 47 | 73 | |
GenBank mRNA | BC134791.1 | - | 1 - 2 | 60 | 73 | |
X97635.1 | - | 47 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
